3-(2-Fluoroethoxy)phenylboronic acid 96%

Supplier: Apollo Scientific

Synonyms: (3-(2-Fluoroethoxy)phenyl)boronic acid

PC901356-1G PC901356-250MG
APOSPC901356-1GEA 337 GBP
APOSPC901356-1G APOSPC901356-250MG
3-(2-Fluoroethoxy)phenylboronic acid 96%
3-(2-Fluoroethoxy)phenylboronic acid
Formula: C₈H₁₀BFO₃
MW: 183.973 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18783120
CAS Number: 2096332-11-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR