2,3-Difluoro-5-methylphenylboronic acid 98%
Supplier: Apollo Scientific
PC901270-10G
PC901270-1G
PC901270-5G
APOSPC901270-10GEA
453
GBP
APOSPC901270-10G
APOSPC901270-1G
APOSPC901270-5G
2,3-Difluoro-5-methylphenylboronic acid 98%
2,3-Difluoro-5-methylphenylboronic acid
Formula:
C₇H₇BF₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD22573057 CAS Number: 934247-79-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...