1-Chloro-4-fluoro-2-methyl-5-nitrobenzene 97%
Supplier: Apollo Scientific
PC900714-1G
PC900714-5G
APOSPC900714-1GEA
70.9
GBP
APOSPC900714-1G
APOSPC900714-5G
1-Chloro-4-fluoro-2-methyl-5-nitrobenzene 97%
1-Chloro-4-fluoro-2-methyl-5-nitrobenzene
Formula:
C₇H₅ClFNO₂ MW: 189,572 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11110548 CAS Number: 170098-88-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...