2-Chloro-6-(trifluoromethyl)benzaldehyde 95+%

Supplier: Apollo Scientific
Warning

Synonyms: 3-Chloro-2-formylbenzotrifluoride, 2-Chloro-6-trifluoromethylbenzaldehyde, 6-Chloro-α,α,α-trifluoro-o-tolualdehyde

PC8832-1G PC8832-25G PC8832-5G
APOSPC8832-1GEA 14.5 GBP
APOSPC8832-1G APOSPC8832-25G APOSPC8832-5G
2-Chloro-6-(trifluoromethyl)benzaldehyde 95+%
2-Chloro-6-(trifluoromethyl)benzaldehyde
Formula: C₈H₄ClF₃O
MW: 208,565 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06660231
CAS Number: 60611-22-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR