6-Fluoro-5-methyl-1H-indole 98%

Supplier: Apollo Scientific

PC8302-1G PC8302-250MG PC8302-5G
APOSPC8302-1GEA 326 GBP
APOSPC8302-1G APOSPC8302-250MG APOSPC8302-5G
6-Fluoro-5-methyl-1H-indole 98%
6-Fluoro-5-methyl-1H-indole
Formula: C₉H₈FN
MW: 149,165 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03094489
CAS Number: 162100-95-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR