5,5,5-Trifluoro-DL-norvaline 96%

Supplier: Apollo Scientific
Warning

Synonyms: 2-Amino-5,5,5-trifluorovaleric acid, 2-Amino-5,5,5-trifluoropentanoic acid

PC7813-100MG PC7813-1G PC7813-250MG
APOSPC7813-100MGEA 98.3 GBP
APOSPC7813-100MG APOSPC7813-1G APOSPC7813-250MG
5,5,5-Trifluoro-DL-norvaline 96%
5,5,5-Trifluoro-DL-norvaline
Formula: C₅H₈F₃NO₂
MW: 171,118 g/mol
Melting Pt: 256…258 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00190648
CAS Number: 2365-80-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR