4-Bromo-2-fluorobenzaldoxime 98%

Supplier: Apollo Scientific
Warning

PC7368-1G PC7368-5G
APOSPC7368-1GEA 163 GBP
APOSPC7368-1G APOSPC7368-5G
4-Bromo-2-fluorobenzaldoxime 98%
4-Bromo-2-fluorobenzaldoxime
Formula: C₇H₅BrFNO
MW: 218.023 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00143262
CAS Number: 202865-64-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR