1,1,3-Trichlorotrifluoroacetone 97%

Supplier: Apollo Scientific
Danger

PC7040-25G PC7040-5G
APOSPC7040-25GEA 103 GBP
APOSPC7040-25G APOSPC7040-5G
1,1,3-Trichlorotrifluoroacetone 97%
1,1,3-Trichlorotrifluoroacetone
Formula: C₃Cl₃F₃O
MW: 215,386 g/mol
Boiling Pt: 84,5 °C (760 mm Hg)
Melting Pt: 44415 °C
Storage Temperature: Ambient
MDL Number: MFCD00039321
CAS Number: 79-52-7
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR