(2Z)-2,4,4,4-Tetrafluorobut-2-en-1-ol 97+%

Supplier: Apollo Scientific
Warning

Synonyms: (2Z)-1-Hydroxy-2,4,4,4-tetrafluorobut-2-ene

PC6428-1G PC6428-250MG
APOSPC6428-1GEA 302 GBP
APOSPC6428-1G APOSPC6428-250MG
(2Z)-2,4,4,4-Tetrafluorobut-2-en-1-ol 97+%
(2Z)-2,4,4,4-Tetrafluorobut-2-en-1-ol
Formula: C₄H₄F₄O
MW: 144,068 g/mol
Boiling Pt: 76…78 °C (28 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD07784255
CAS Number: 91600-37-2
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR