3-[(3-Fluorophenyl)amino]propanoic acid
Supplier: Apollo Scientific
Synonyms:
3-[(3-Fluorophenyl)amino]propionic acid, 3-((3-Fluorophenyl)amino)propanoic acid
PC6367-1G
PC6367-5G
APOSPC6367-1GEA
112
GBP
APOSPC6367-1G
APOSPC6367-5G
3-[(3-Fluorophenyl)amino]propanoic acid
3-[(3-Fluorophenyl)amino]propanoic acid
Formula:
C₉H₁₀FNO₂ MW: 183,18 g/mol Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD07784303 CAS Number: 885275-89-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...