1-(Perfluorohex-1-yl)decane 95%

Supplier: Apollo Scientific
Warning

Synonyms: 1,1,1,2,2,3,3,4,4,5,5,6,6-Tridecafluorohexadecane

PC6086B-5G PC6086B-1G
APOSPC6086B-5GEA 81.2 GBP
APOSPC6086B-5G APOSPC6086B-1G
1-(Perfluorohex-1-yl)decane 95%
1-(Perfluorohex-1-yl)decane
Formula: C₁₆H₂₁F₁₃
MW: 460.317 g/mol
Boiling Pt: 105…106 °C (3 mm Hg)
Density: 1.291 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00236704
CAS Number: 147492-59-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR