6H-Perfluorohex-1-ene 97%
Supplier: Apollo Scientific
|
Warning
|
PC6085E-1G
PC6085E-5G
APOSPC6085E-1GEA
132
GBP
APOSPC6085E-1G
APOSPC6085E-5G
6H-Perfluorohex-1-ene 97%
6H-Perfluorohex-1-ene
Formula:
C₆HF₁₁ MW: 282,055 g/mol Boiling Pt: 74…75 °C (760 mm Hg) Density: 1,654 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00156093 CAS Number: 1767-94-8 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...