1H-Perfluorooct-1-yne

Supplier: Apollo Scientific
Warning

Synonyms: 3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluorooct-1-yne

PC5985-1G PC5985-5G
APOSPC5985-1GEA 283 GBP
APOSPC5985-1G APOSPC5985-5G
1H-Perfluorooct-1-yne
1H-Perfluorooct-1-yne
Formula: C₈HF₁₃
MW: 344,073 g/mol
Boiling Pt: 95 °C
Storage Temperature: Ambient
MDL Number: MFCD00042347
CAS Number: 55756-24-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR