2H-Perfluoroisopropyl benzoate 97%

Supplier: Apollo Scientific

Synonyms: 1,1,13,3,3-Hexafluoroisopropyl benzoate

PC5862-1G PC5862-5G
APOSPC5862-1GEA 87.8 GBP
APOSPC5862-1G APOSPC5862-5G
2H-Perfluoroisopropyl benzoate 97%
2H-Perfluoroisopropyl benzoate
Formula: C₁₀H₆F₆O₂
MW: 272,144 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00769374
CAS Number: 10315-85-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR