8-Bromo-1-fluorophenazine 94%

Supplier: Apollo Scientific
Warning

Synonyms: 3-Bromo-6-fluorophenazine

PC57775-1G PC57775-250MG PC57775-5G
APOSPC57775-1GEA 81.2 GBP
APOSPC57775-1G APOSPC57775-250MG APOSPC57775-5G
8-Bromo-1-fluorophenazine 94%
8-Bromo-1-fluorophenazine
Formula: C₁₂H₆BrFN₂
MW: 277.092 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32070710
CAS Number: 2379918-33-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR