2,3,3-Trifluoropropene

Supplier: Apollo Scientific
Danger

Synonyms: 1243yf

PC5756-10G PC5756-5G
APOSPC5756-10GEA 591 GBP
APOSPC5756-10G APOSPC5756-5G
2,3,3-Trifluoropropene
2,3,3-Trifluoropropene
Formula: C₃H₃F₃
MW: 96,051 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10565608
CAS Number: 158664-13-2
UN: 1954
ADR: 2.1,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR