3-Bromoperfluoroprop-1-ene

Supplier: Apollo Scientific
Warning

Synonyms: 3-Bromo-1,1,2,3,3-pentafluoroprop-1-ene

PC5705-1G PC5705-5G
APOSPC5705-1GEA 178 GBP
APOSPC5705-1G APOSPC5705-5G
3-Bromoperfluoroprop-1-ene
3-Bromoperfluoroprop-1-ene
Formula: C₃BrF₅
MW: 210,928 g/mol
Boiling Pt: 25,5 °C (738 mm Hg)
Melting Pt: –165 °C
Density: 1,812 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD07784211
CAS Number: 431-56-1
UN: 2810
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR