Amino(4-fluorophenyl)acetonitrile hydrochloride 97%

Supplier: Apollo Scientific
Warning

PC57027-1G PC57027-5G
APOSPC57027-1GEA 138 GBP
APOSPC57027-1G APOSPC57027-5G
Amino(4-fluorophenyl)acetonitrile hydrochloride 97%
Amino(4-fluorophenyl)acetonitrile hydrochloride
Formula: C₈H₇FN₂·ClH
Storage Temperature: Ambient
MDL Number: MFCD20489386
CAS Number: 934830-01-0
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR