5-Bromo-7-fluorobenzofuran-2-carboxaldehyde 95%

Supplier: Apollo Scientific
Warning

PC56924-1G PC56924-250MG
APOSPC56924-1GEA 420 GBP
APOSPC56924-1G APOSPC56924-250MG
5-Bromo-7-fluorobenzofuran-2-carboxaldehyde 95%
5-Bromo-7-fluorobenzofuran-2-carboxaldehyde
Formula: C₉H₄BrFO₂
MW: 243,029 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21233546
CAS Number: 1480288-05-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR