2-Bromo-6-fluoro-3-(trifluoromethyl)phenacyl bromide 95%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
2-Bromo-1-[2-bromo-6-fluoro-3-(trifluoromethyl)phenyl]ethanone
PC56705-100MG
PC56705-1G
PC56705-250MG
APOSPC56705-100MGEA
129
GBP
APOSPC56705-100MG
APOSPC56705-1G
APOSPC56705-250MG
2-Bromo-6-fluoro-3-(trifluoromethyl)phenacyl bromide 95%
2-Bromo-6-fluoro-3-(trifluoromethyl)phenacyl bromide
Formula:
C₉H₄Br₂F₄O MW: 363,929 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD31618859 CAS Number: 2167600-04-4 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...