2-Fluoro-4-(methylthio)pyridine

Supplier: Apollo Scientific
Warning

PC56138-1G PC56138-250MG
APOSPC56138-1GEA 392 GBP
APOSPC56138-1G APOSPC56138-250MG
2-Fluoro-4-(methylthio)pyridine
2-Fluoro-4-(methylthio)pyridine
Formula: C₆H₆FNS
MW: 143,182 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23713127
CAS Number: 1428234-71-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR