4-Bromo-2,5-difluorobenzyl alcohol

Supplier: Apollo Scientific
Warning

Synonyms: (4-Bromo-2,5-difluorophenyl)methanol

PC56037-1G PC56037-5G PC56037-250MG
APOSPC56037-1GEA 182 GBP
APOSPC56037-1G APOSPC56037-5G APOSPC56037-250MG
4-Bromo-2,5-difluorobenzyl alcohol
4-Bromo-2,5-difluorobenzyl alcohol
Formula: C₇H₅BrF₂O
MW: 223,015 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11847046
CAS Number: 486460-26-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR