3'-Bromo-4'-iodo-2,2,2-trifluoroacetophenone

Supplier: Apollo Scientific
Warning

Synonyms: 1-(3-Bromo-4-iodophenyl)-2,2,2-trifluoroethanone

PC53430-1G PC53430-250MG
APOSPC53430-1GEA 505 GBP
APOSPC53430-1G APOSPC53430-250MG
3'-Bromo-4'-iodo-2,2,2-trifluoroacetophenone
3'-Bromo-4'-iodo-2,2,2-trifluoroacetophenone
Formula: C₈H₃BrF₃IO
Storage Temperature: Ambient
MDL Number: MFCD30481129
CAS Number: 2149598-84-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR