4'-(Difluoromethyl)acetophenone
Supplier: Apollo Scientific
|
Warning
|
PC53336-1G
PC53336-250MG
APOSPC53336-1GEA
231
GBP
APOSPC53336-1G
APOSPC53336-250MG
4'-(Difluoromethyl)acetophenone
4'-(Difluoromethyl)acetophenone
Formula:
C₉H₈F₂O Storage Temperature: Ambient |
MDL Number:
MFCD11042622 CAS Number: 179990-93-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...