5-Amino-3-(trifluoromethyl)picolinonitrile 97%
Supplier: Apollo Scientific
|
Danger
|
PC53326-1G
PC53326-5G
PC53326-10G
APOSPC53326-1GEA
106
GBP
APOSPC53326-1G
APOSPC53326-5G
APOSPC53326-10G
5-Amino-3-(trifluoromethyl)picolinonitrile 97%
5-Amino-3-(trifluoromethyl)picolinonitrile
Formula:
C₇H₄F₃N₃ MW: 187,122 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD17010171 CAS Number: 573762-62-6 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...