6-Fluoro-2,3-dihydro-1H-indole
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
6-Fluoroindoline
PC53293-100MG
PC53293-250MG
APOSPC53293-100MGEA
59.1
GBP
APOSPC53293-100MG
APOSPC53293-250MG
6-Fluoro-2,3-dihydro-1H-indole
6-Fluoro-2,3-dihydro-1H-indole
Formula:
C₈H₈FN MW: 137,154 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00022791 CAS Number: 2343-23-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...