2,4-Difluoro-5-(trifluoromethyl)benzyl alcohol

Supplier: Apollo Scientific
Warning

Synonyms: [2,4-Difluoro-5-(trifluoromethyl)phenyl]methanol

PC53037-1G PC53037-250MG
APOSPC53037-1GEA 211 GBP
APOSPC53037-1G APOSPC53037-250MG
2,4-Difluoro-5-(trifluoromethyl)benzyl alcohol
2,4-Difluoro-5-(trifluoromethyl)benzyl alcohol
Formula: C₈H₅F₅O
MW: 212,117 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28897197
CAS Number: 1936257-01-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR