3-Amino-5-bromo-4-hydroxyphenylsulphur pentafluoride 95%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
3-Bromo-2-hydroxy-5-(pentafluorosulphanyl)aniline, 2-Amino-6-bromo-4-(pentafluorosulphanyl)phenol
PC5248-1G
PC5248-250MG
APOSPC5248-1GEA
322
GBP
APOSPC5248-1G
APOSPC5248-250MG
3-Amino-5-bromo-4-hydroxyphenylsulphur pentafluoride 95%
3-Amino-5-bromo-4-hydroxyphenylsulphur pentafluoride
Formula:
C₆H₅BrF₅NOS Storage Temperature: Ambient |
MDL Number:
MFCD11501042 CAS Number: 1159512-29-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...