3-Bromo-5-iodobenzal fluoride

Supplier: Apollo Scientific
Warning

PC52190-1G PC52190-250MG PC52190-5G
APOSPC52190-1GEA 519 GBP
APOSPC52190-1G APOSPC52190-250MG APOSPC52190-5G
3-Bromo-5-iodobenzal fluoride
3-Bromo-5-iodobenzal fluoride
Formula: C₇H₄BrF₂I
Storage Temperature: Ambient
MDL Number: MFCD18391111
CAS Number: 1261476-63-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR