2-Bromo-4-chloro-1-(trifluoromethoxy)benzene 95%
Supplier: Apollo Scientific
PC520646-1G
PC520646-250MG
APOSPC520646-1GEA
95.7
GBP
APOSPC520646-1G
APOSPC520646-250MG
2-Bromo-4-chloro-1-(trifluoromethoxy)benzene 95%
2-Bromo-4-chloro-1-(trifluoromethoxy)benzene
Formula:
C₇H₃BrClF₃O MW: 275,45 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11847191 CAS Number: 1260810-00-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...