Bis-(3-Bromo-2-fluorobenzyl)ether 97%
Supplier: Apollo Scientific
PC520635-1G
PC520635-5G
APOSPC520635-1GEA
227
GBP
APOSPC520635-1G
APOSPC520635-5G
Bis-(3-Bromo-2-fluorobenzyl)ether 97%
Bis-(3-Bromo-2-fluorobenzyl)ether
Formula:
C₁₄H₁₀Br₂F₂O MW: 392.033 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11226505 CAS Number: 1186195-12-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...