2,4-Difluoro-5-nitrobenzyl bromide 95%

Supplier: Apollo Scientific
Danger

Synonyms: 1-(Bromomethyl)-2,4-difluoro-5-nitrobenzene

PC51226-1G PC51226-250MG PC51226-5G
APOSPC51226-1GEA 441 GBP
APOSPC51226-1G APOSPC51226-250MG APOSPC51226-5G
2,4-Difluoro-5-nitrobenzyl bromide 95%
2,4-Difluoro-5-nitrobenzyl bromide
Formula: C₇H₄BrF₂NO₂
MW: 252,013 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12756738
CAS Number: 1260790-43-9
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR