2-Bromo-4-(trifluoromethoxy)phenylacetylene

Supplier: Apollo Scientific
Warning

Synonyms: 2-Bromo-1-ethynyl-4-(trifluoromethoxy)benzene, 3-Bromo-4-ethynylphenyl trifluoromethyl ether

PC51142-1G PC51142-250MG
APOSPC51142-1GEA 132 GBP
APOSPC51142-1G APOSPC51142-250MG
2-Bromo-4-(trifluoromethoxy)phenylacetylene
2-Bromo-4-(trifluoromethoxy)phenylacetylene
Formula: C₉H₄BrF₃O
MW: 265,027 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD23166760
CAS Number: 1426290-11-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR