3-(Dimethylamino)-4-fluorobenzeneboronic acid

Supplier: Apollo Scientific

Synonyms: (3-(Dimethylamino)-4-fluorophenyl)boronic acid

PC510348-1G PC510348-5G PC510348-250MG
APOSPC510348-1GEA 358 GBP
APOSPC510348-1G APOSPC510348-5G APOSPC510348-250MG
3-(Dimethylamino)-4-fluorobenzeneboronic acid
3-(Dimethylamino)-4-fluorobenzeneboronic acid
Formula: C₈H₁₁BFNO₂
MW: 182,988 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22573089
CAS Number: 1186215-34-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR