6,8-Difluorochroman-4-one

Supplier: Apollo Scientific
Warning

PC510240-1G PC510240-250MG PC510240-5G
APOSPC510240-1GEA 387 GBP
APOSPC510240-1G APOSPC510240-250MG APOSPC510240-5G
6,8-Difluorochroman-4-one
6,8-Difluorochroman-4-one
Formula: C₉H₆F₂O₂
MW: 184,14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01764784
CAS Number: 259655-01-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR