6-Bromo-5-fluoro-3-iodo-1H-indazole
Supplier: Apollo Scientific
|
Warning
|
PC51019-1G
PC51019-100MG
PC51019-500MG
APOSPC51019-1GEA
512
GBP
APOSPC51019-1G
APOSPC51019-100MG
APOSPC51019-500MG
6-Bromo-5-fluoro-3-iodo-1H-indazole
6-Bromo-5-fluoro-3-iodo-1H-indazole
Formula:
C₇H₃BrFIN₂ MW: 340,919 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD18384871 CAS Number: 1286734-77-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...