3-Bromo-5-chloro-2-fluorobenzaldehyde 95%

Supplier: Apollo Scientific

PC50566-1G PC50566-5G PC50566-25G
APOSPC50566-1GEA 205 GBP
APOSPC50566-1G APOSPC50566-5G APOSPC50566-25G
3-Bromo-5-chloro-2-fluorobenzaldehyde 95%
3-Bromo-5-chloro-2-fluorobenzaldehyde
Formula: C₇H₃BrClFO
MW: 237,454 g/mol
Melting Pt: 59…60 °C
Storage Temperature: Ambient
MDL Number: MFCD22627836
CAS Number: 1269440-82-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR