3-Bromo-4-(trifluoromethoxy)phenylisocyanate
Supplier: Apollo Scientific
|
Danger
|
PC503008-1G
PC503008-25G
PC503008-5G
APOSPC503008-1GEA
19.7
GBP
APOSPC503008-1G
APOSPC503008-25G
APOSPC503008-5G
3-Bromo-4-(trifluoromethoxy)phenylisocyanate
3-Bromo-4-(trifluoromethoxy)phenylisocyanate
May form small amount of sediment
Formula:
C₈H₃BrF₃NO₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD32200431 CAS Number: 685859-17-0 UN: 2206 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...