3-Bromo-5-methylbenzyl fluoride 95%

Supplier: Apollo Scientific
Danger

PC502884-1G PC502884-250MG
APOSPC502884-1GEA 551 GBP
APOSPC502884-1G APOSPC502884-250MG
3-Bromo-5-methylbenzyl fluoride 95%
3-Bromo-5-methylbenzyl fluoride
Formula: C₈H₈BrF
MW: 203,052 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31634829
CAS Number: 2244087-79-2
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR