Methyl 3-fluoro-5-iodobenzoate 95%
Supplier: Apollo Scientific
PC502587-1G
PC502587-250MG
APOSPC502587-1GEA
484
GBP
APOSPC502587-1G
APOSPC502587-250MG
Methyl 3-fluoro-5-iodobenzoate 95%
Methyl 3-fluoro-5-iodobenzoate
Formula:
C₈H₆FIO₂ MW: 280,035 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18399306 CAS Number: 1261478-09-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...