2-Bromo-3',4'-difluoropropiophenone
Supplier: Apollo Scientific
|
Danger
|
PC502341-1G
PC502341-250MG
PC502341-5G
APOSPC502341-1GEA
398
GBP
APOSPC502341-1G
APOSPC502341-250MG
APOSPC502341-5G
2-Bromo-3',4'-difluoropropiophenone
2-Bromo-3',4'-difluoropropiophenone
Formula:
C₉H₇BrF₂O MW: 249,052 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11182355 CAS Number: 23384-75-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...