3,5-Difluoro-2-nitrobenzonitrile 95%

Supplier: Apollo Scientific
Warning

PC502143-1G PC502143-250MG
APOSPC502143-1GEA 107 GBP
APOSPC502143-1G APOSPC502143-250MG
3,5-Difluoro-2-nitrobenzonitrile 95%
3,5-Difluoro-2-nitrobenzonitrile
Formula: C₇H₂F₂N₂O₂
MW: 184,1 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13185568
CAS Number: 868596-13-8
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR