2-Bromo-2',6'-difluoropropiophenone
Supplier: Apollo Scientific
|
Danger
|
PC502135-1G
PC502135-250MG
PC502135-5G
APOSPC502135-1GEA
321
GBP
APOSPC502135-1G
APOSPC502135-250MG
APOSPC502135-5G
2-Bromo-2',6'-difluoropropiophenone
2-Bromo-2',6'-difluoropropiophenone
Formula:
C₉H₇BrF₂O MW: 249,052 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18391236 CAS Number: 481066-21-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...