Methyl 2-fluoro-3-formylbenzoate 98%

Supplier: Apollo Scientific

PC501624-1G PC501624-5G
APOSPC501624-1GEA 347 GBP
APOSPC501624-1G APOSPC501624-5G
Methyl 2-fluoro-3-formylbenzoate 98%
Methyl 2-fluoro-3-formylbenzoate
Formula: C₉H₇FO₃
MW: 182,148 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24447977
CAS Number: 1262419-96-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR