3-Fluoro-2-nitrophenacyl bromide 95%

Supplier: Apollo Scientific
Danger

PC501205-1G PC501205-250MG PC501205-5G
APOSPC501205-1GEA 472 GBP
APOSPC501205-1G APOSPC501205-250MG APOSPC501205-5G
3-Fluoro-2-nitrophenacyl bromide 95%
3-Fluoro-2-nitrophenacyl bromide
Formula: C₈H₅BrFNO₃
MW: 262,033 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23711759
CAS Number: 1427431-58-0
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR