4-Fluoro-3-methylbenzene-1,2-diamine 95%
Supplier: Apollo Scientific
|
Warning
|
PC500346-1G
PC500346-250MG
APOSPC500346-1GEA
317
GBP
APOSPC500346-1G
APOSPC500346-250MG
4-Fluoro-3-methylbenzene-1,2-diamine 95%
4-Fluoro-3-methylbenzene-1,2-diamine
Formula:
C₇H₉FN₂ MW: 140,158 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09026959 CAS Number: 485832-95-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...