4-Bromo-2-fluoro-5-nitrophenol 95%

Supplier: Apollo Scientific
Warning

PC500262-1G PC500262-250MG
APOSPC500262-1GEA 231 GBP
APOSPC500262-1G APOSPC500262-250MG
4-Bromo-2-fluoro-5-nitrophenol 95%
4-Bromo-2-fluoro-5-nitrophenol
Formula: C₆H₃BrFNO₃
MW: 235,995 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11100973
CAS Number: 661463-12-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR