3-Bromo-2-(trifluoromethyl)phenol 96%
Supplier: Apollo Scientific
PC499003-1G
PC499003-25G
PC499003-5G
APOSPC499003-1GEA
130
GBP
APOSPC499003-1G
APOSPC499003-25G
APOSPC499003-5G
3-Bromo-2-(trifluoromethyl)phenol 96%
3-Bromo-2-(trifluoromethyl)phenol
Formula:
C₇H₄BrF₃O MW: 241,005 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD13194489 CAS Number: 1214352-26-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...