Perfluoro[(tert-butyl)cyclohexane]

Supplier: Apollo Scientific
Warning

Synonyms: [1,1,1,3,3,3-Hexafluoro-2-(trifluoromethyl)prop-2-yl](undecafluoro)cyclohexane, Icosafluoro[(tert-butyl)cyclohexane]

PC49620-1G PC49620-25G PC49620-5G
APOSPC49620-1GEA 51.2 GBP
APOSPC49620-1G APOSPC49620-25G APOSPC49620-5G
Perfluoro[(tert-butyl)cyclohexane]
Perfluoro[(tert-butyl)cyclohexane]
Formula: C₁₀F₂₀
MW: 500,075 g/mol
Boiling Pt: 148,5 °C
Melting Pt: –24 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00153230
CAS Number: 84808-64-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR