Potassium pentafluorobenzoate 95%

Supplier: Apollo Scientific

PC49473-1G PC49473-25G PC49473-5G PC49473-10G
APOSPC49473-1GEA 57.7 GBP
APOSPC49473-1G APOSPC49473-25G APOSPC49473-5G APOSPC49473-10G
Potassium pentafluorobenzoate 95%
Potassium pentafluorobenzoate
Formula: C₇F₅O₂·K
MW: 250,164 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09752634
CAS Number: 58521-27-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR